Order Entry
Switzerland
ContactUsLinkComponent
Methanol, HiPerSolv CHROMANORM®, Ultra Plus for LC-MS, suitable for UPLC/UHPLC-MS instruments
Flammable
Toxic
Health
Methanol, HiPerSolv CHROMANORM®, Ultra Plus for LC-MS, suitable for UPLC/UHPLC-MS instruments
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Toxic
Health
Methanol, HiPerSolv CHROMANORM®, Ultra Plus for LC-MS, suitable for UPLC/UHPLC-MS instruments
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Appearance:
    Clear colorless liquid
  • Color (APHA, Pt‑Co):
    max. 5
  • Assay (GC, on anhydrous basis):
    min. 99.98 %
  • Residue after evaporation:
    max. 0.0001 %w/w
  • Water (KF):
    max. 0.03 %w/w
  • Acidity (as Acetic acid):
    max. 0.002 %
  • Alkalinity (as Ammonia):
    max. 0.0001 %
  • UHPLC/MS/ESI+ (as Reserpine):
    max. 6 ppb
  • H.Peak by PDAD 220‑400nm:
    max. 0.004 AU
  • Grad. elution H.Peak at 220nm:
    max. 0.004 AU
  • Grad. elution drift at 220nm:
    max. 0.05 AU
  • Grad. elution H.Peak at 235nm:
    max. 0.002 AU
  • Grad. elution drift at 235nm:
    max. 0.015 AU
  • F254nm (as Quinine):
    max. 0.5 ppb
  • F365nm (as Quinine):
    max. 0.3 ppb
  • T210nm:
    min. 40 %
  • T220nm:
    min. 65 %
  • T230nm:
    min. 80 %
  • T260nm:
    min. 98 %
  • Ag (Silver):
    max. 50 ppb
  • Al (Aluminum):
    max. 20 ppb
  • Ba (Barium):
    max. 50 ppb
  • Bi (Bismuth):
    max. 50 ppb
  • Ca (Calcium):
    max. 50 ppb
  • Cd (Cadmium):
    max. 50 ppb
  • Co (Cobalt):
    max. 20 ppb
  • Cr (Chromium):
    max. 20 ppb
  • Cu (Copper):
    max. 20 ppb
  • Fe (Iron):
    max. 20 ppb
  • K (Potassium):
    max. 50 ppb
  • Li (Lithium):
    max. 50 ppb
  • Mg (Magnesium):
    max. 20 ppb
  • Mn (Manganese):
    max. 20 ppb
  • Mo (Molybdenum):
    max. 50 ppb
  • Na (Sodium):
    max. 50 ppb
  • Ni (Nickel):
    max. 20 ppb
  • Pb (lead):
    max. 20 ppb
  • Sn (Tin):
    max. 50 ppb
  • Sr (Strontium):
    max. 50 ppb
  • Zn (Zinc):
    max. 50 ppb

Specification Test Results

Frequently Bought Together