2-Phenylaniline
Catalog # EHERC10173020
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-Aminobiphenyl, Biphenyl-2-ylamine, 2-Biphenylylamine
:
:169.23 g/mol
Boiling Pt:299 °C (1013 hPa)
Melting Pt:42…44 °C
EINECS:201-990-9
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:90-41-5
Formula:C₁₂H₁₁N
MDL Number:MFCD00007701