2,3',4',5-Tetrachlorobiphenyl (PCB No. 70)
Catalog # EHERC20007000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:291.99 g/mol
CAS Number:32598-11-1
Formula:C₁₂H₆Cl₄