Order Entry
Switzerland
ContactUsLinkComponent
2-Mesitylenesulphonyl chloride for synthesis
Corrosive
2-Mesitylenesulphonyl chloride for synthesis
Catalog # 8.14055.0025
Supplier:  Merck
CAS Number:  
Corrosive
2-Mesitylenesulphonyl chloride for synthesis
Catalog # 8.14055.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    53 - 56 °C
  • Molar Mass:
    218.70 g/mol
  • Flash Point:
    113 °C (External MSDS)

Specification Test Results

About this item

 :
 :218.7 g/mol
Boiling Pt:150 °C (20 mmHg)
Melting Pt:53…56 °C
Storage Temperature:Ambient
EINECS:212-257-8
UN:3261
ADR:8,II
 :GHS05
CAS Number:773-64-8
Formula:C₉H₁₁ClO₂S
MDL Number:MFCD00007434