Order Entry
Switzerland
ContactUsLinkComponent
2-Mesitylenesulphonyl chloride 99%
Corrosive
2-Mesitylenesulphonyl chloride 99%
Certificates
undefined
Corrosive
2-Mesitylenesulphonyl chloride 99%

Specification Test Results

  • Appearance:
    White to grey Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    53 to 57 °C
  • Titration Argentometric:
    ≥98.5 % (ex Cl)

Specification Test Results

Related Information

Frequently Bought Together