VWR® TraceableGO, Temperature and Humidity Data Loggers with Bluetooth
About this item
TraceableGO Bluetooth temperature and humidity dataloggers fill in the unknowns between destinations. Bluetooth device is the ideal transit partner for monitoring materials between destinations. Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location.
- Compact and easy to use
- Critical information (temperature, trip duration, MIN/MAX, battery level and alarms) all displayed on LCD at all times
- Compatible with TraceableGO or TraceableLIVE mobile apps
- Devices store up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours, logging duration is 227 days (7,5 months)
- TraceableGO bluetooth devices are pre-programmed with alarm values
How does it work:
Press START to begin trip monitoring
Place device inside a small cooler or even an entire pallet of material
Once the trip has ended, double-press START/STOP to enable Bluetooth and export the data using the TraceableGO™ mobile app
TraceableGO mobile app immediately identifies if the contents experienced conditions outside user-defined parameters
Certifications: Multi-point calibration on an individually-numbered Traceable certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM mutual recognition agreement. Supplied with battery and Traceable certificate.
2 Options Available Below
Product Details & Documents
TraceableGO Bluetooth temperature and humidity dataloggers fill in the unknowns between destinations. Bluetooth device is the ideal transit partner for monitoring materials between destinations. Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location.
- Compact and easy to use
- Critical information (temperature, trip duration, MIN/MAX, battery level and alarms) all displayed on LCD at all times
- Compatible with TraceableGO or TraceableLIVE mobile apps
- Devices store up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours, logging duration is 227 days (7,5 months)
- TraceableGO bluetooth devices are pre-programmed with alarm values
How does it work:
Press START to begin trip monitoring
Place device inside a small cooler or even an entire pallet of material
Once the trip has ended, double-press START/STOP to enable Bluetooth and export the data using the TraceableGO™ mobile app
TraceableGO mobile app immediately identifies if the contents experienced conditions outside user-defined parameters
Certifications: Multi-point calibration on an individually-numbered Traceable certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM mutual recognition agreement. Supplied with battery and Traceable certificate.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 620-2673
- 620-2674
- Alarm:
- 90 alarm levels
- 90 alarm levels
- Display:
- LCD
- LCD
- Interface:
- Bluetooth
- Bluetooth
- Recording intervals:
- 1 min - 12 hr
- 1 min - 12 hr
- Weight:
- 19 g
- 19 g
- W×D×H:
- 89×20×51 mm
- 89×20×51 mm
- Data memory - No. of readings:
- 65536
- 65536
- Batteries:
- 2×AAA alkaline batteries, 3 V
- 2×AAA alkaline batteries, 3 V
Specifications
Product Family Options
Product Information
- RangeAccuracyResolutionAvailabilityPrice
- 620-2673Temperature logger, TraceableGO, bluetooth, 6535Temperature logger, TraceableGO, bluetooth, 6535T: ‒20...+70 °C±0,4 °CT: 0,1 °CLog in for availability1 * 1 STSFr. 103.00Specifications:
More
Specifications
Cat. No.620-2673Alarm90 alarm levelsDisplayLCDInterfaceBluetoothRecording intervals1 min - 12 hrWeight19 gW×D×H89×20×51 mmData memory - No. of readings65536Batteries2×AAA alkaline batteries, 3 V - 620-2674Temperature and humidity logger, TraceableGO, bluetooth, 6537Temperature and humidity logger, TraceableGO, bluetooth, 6537T: ‒20...+70 °C
RH: 0 - 95 %T: ±0,4 °C (–10...+70 °C), otherwise ±0,5 °C
RH: ±3% (5 - 75%), otherwise ±5 %T: 0,1 °C
RH: 0,1 %Log in for availability1 * 1 STSFr. 122.00Specifications:More
Specifications
Cat. No.620-2674Alarm90 alarm levelsDisplayLCDInterfaceBluetoothRecording intervals1 min - 12 hrWeight19 gW×D×H89×20×51 mmData memory - No. of readings65536Batteries2×AAA alkaline batteries, 3 V
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



