Order Entry
Switzerland
ContactUsLinkComponent
Phenol red ACS
Phenol red ACS
Supplier:  Merck
Certificates
undefined
Phenol red ACS
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    0.77 g/l (100 °C)
  • Melting Point:
    300 °C
  • Molar Mass:
    354.38 g/mol
  • Bulk Density:
    200 - 300 kg/m3

Specification Test Results

Related Information

Frequently Bought Together