
Phenolphthalein ACS, Reag. Ph. Eur., Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:77-09-8
:
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913
:318.33 g/mol
Merck Index:14,07243
Melting Pt:258…262 °C
Density:1.3 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:201-004-7
REACH:01-2119498295-24
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:3.36 mg/l (20 °C)
- Melting Point:263.7 °C
- Molar Mass:318.32 g/mol
- Bulk Density:350 - 450 kg/m3
- Density:1.296 g/cm3 (20 °C)
- pH:- 0.00001 Pa (50 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.07233.0025_TDS_EN ProductPage_107233
Product specification
msds_107233
SDS
Frequently Bought Together