ContactUsLinkComponent
Phenolphthalein ACS
Health
Phenolphthalein ACS
Certificates
undefined
Health
Phenolphthalein ACS
Certificates
Specification Test Results
  • Appearance:
    Off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Visual transition interval:
    From pH 8.0 (colorless) to pH 10 (red)
  • Clarity of alcohol solution:
    Passes test