Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bromothymol blue

Bromothymol blue

Glass bottle for solids, 5 g
Catalog # ROTHT117.1
Marketsource
Supplier:   Roth Carl
Bromothymol blue
Catalog # ROTHT117.1
Supplier:  Roth Carl
Marketsource
CAS Number:  76-59-5
Bromothymol blue

Glass bottle for solids, 5 g

CAS Number:  76-59-5
Price
SFr. 35.90
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-59-5
  • Formula:C₂₇H₂₈Br₂O₅S
  • MDL Number:MFCD00005872
  • Molecular Weight:624.39 g/mol
  • Merck Index:13,01430
  • Melting Pt:204…202 °C
  • Storage Temperature:Ambient
  • EINECS:200-971-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.