Bromothymol blue
Glass bottle for solids, 25 g
Catalog # MOLE35458451-25G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-59-5
- Formula:C₂₇H₂₈Br₂O₅S
- MDL Number:MFCD00005872
- Molecular Weight:624.39 g/mol
- Merck Index:13,01430
- Melting Pt:204…202 °C
- Storage Temperature:Ambient
- EINECS:200-971-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



