Bromothymol blue
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 76-59-5 Molecular Formula: C27H28Br2O5S Formula Weight: 624.39 Melting Point: 200-202° Beilstein Reference: 373934 Merck Reference: 14,1445 MDL No.: MFCD00005872
Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-59-5
- Formula:C₂₇H₂₈Br₂O₅S
- MDL Number:MFCD00005872
- Molecular Weight:624.39 g/mol
- Merck Index:14,1445
- Melting Pt:200…202 °C
- Storage Temperature:Ambient
- EINECS:200-971-2
3 Options Available Below
Product Details & Documents
CAS No.: 76-59-5 Molecular Formula: C27H28Br2O5S Formula Weight: 624.39 Melting Point: 200-202° Beilstein Reference: 373934 Merck Reference: 14,1445 MDL No.: MFCD00005872
Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-59-5
- Formula:C₂₇H₂₈Br₂O₅S
- MDL Number:MFCD00005872
- Molecular Weight:624.39 g/mol
- Merck Index:14,1445
- Melting Pt:200…202 °C
- Storage Temperature:Ambient
- EINECS:200-971-2



