Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bromochlorophenol blue, powder

Bromochlorophenol blue, powder

Supplier:   MP Biomedicals
Bromochlorophenol blue, powder
Supplier:  MP Biomedicals
Certificates
Bromochlorophenol blue, powder

3 Options Available Below

Product Details & Documents

Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.

Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.

Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.

Soluble in ethanol (0,1% w/v - clear yellow solution), methanol (40 mg/4 ml), EGME, water, benzene.

Synonyms:  4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromo-6-chlorophenol] S,S-dioxide
  • CAS Number:2553-71-1
  • Formula:C₁₉H₁₀Br₂Cl₂O₅S
  • MDL Number:MFCD00047291
  • Molecular Weight:581.06 g/mol
  • Boiling Pt:603 °C (1013 hPa)
  • Melting Pt:273 °C
  • EINECS:219-861-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 46.60
    SFr. 46.60
    1 * 5 g
       SFr. 46.60

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 113.00
    SFr. 113.00
    1 * 25 g
       SFr. 113.00

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - SFr. 178.00
    SFr. 178.00
    1 * 50 g
       SFr. 178.00