


Toluene ≥99.3%, Laboratory Reagent
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:108-88-3
:
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS07
:GHS08
:GHS02
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.3 %
- Non-volatile matter:Max. 0.005 %
- Water (Karl Fischer):Max. 0.05 %
- Refractive Index:n20/D 1.496 (lit.)
- Autoignition Temperature:997 °F
- Vapor Pressure:26 mmHg ( 25 °C)
Specification Test Results
Frequently Bought Together