Order Entry
Switzerland
ContactUsLinkComponent
p-Cresol ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Corrosive
Toxic
p-Cresol ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Catalog # 61030-25G
CAS Number:  
Corrosive
Toxic
p-Cresol ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Catalog # 61030-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • assay (alkalimetric):
    Min. 99 %
  • identity (IR):
    complying
  • appearance:
    complying
  • Cadmium (Cd):
    Max. 5 ppm
  • APHA:
    Max. 25
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 5 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm

Specification Test Results

About this item

 :
 :108.14 g/mol
Merck Index:13,02604
Boiling Pt:202 °C (1.013 hPa)
Melting Pt:35.5 °C (1.013 hPa)
Density:1.03 g/cm³ (20 °C)
Flash Pt:86.0 °C (1.013 hPa)
Storage Temperature:Ambient
EINECS:203-398-6
UN:3455
ADR:6.1,II
 :GHS05
 :GHS06
CAS Number:106-44-5
Formula:C₇H₈O
MDL Number:MFCD00002376