Order Entry
Switzerland
ContactUsLinkComponent
 
Toxic
Corrosive
p-Cresol, Supelco®
Catalog # 42429-5G-F
Supplier:  Merck
CAS Number:  
undefined
Toxic
Corrosive
p-Cresol, Supelco®
Catalog # 42429-5G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :108.14 g/mol
Merck Index:13,02604
Boiling Pt:201.8 °C (1013 hPa)
Melting Pt:32…34 °C
Density:1.034 g/cm³ (20 °C)
Flash Pt:86 °C
EINECS:203-398-6
UN:3455
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
 :GHS06
 :GHS05
Storage Temperature:Ambient
CAS Number:106-44-5
Formula:C₇H₈O
MDL Number:MFCD00002376