

p-Cresol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-44-5
:
Formula:C₇H₈O
MDL Number:MFCD00002376
:108.14 g/mol
Merck Index:13,02604
Boiling Pt:201.8 °C (1013 hPa)
Melting Pt:32…34 °C
Density:1.034 g/cm³ (20 °C)
EINECS:203-398-6
UN:3455
ADR:6.1 (8),II
:M-Clarity™ quality level = MQ200
Flash Pt:86 °C
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:20 g/l (20 °C)
- Melting Point:32 - 34 °C
- Molar Mass:108.14 g/mol
- Boiling Point:202.5 °C (1013 hPa)
- Vapor Pressure:0.06 hPa (20 °C)
- Flash Point:86 °C
- Explosion Limit:1 %(V)
- Density:1.034 g/cm3 (20 °C)
- Ignition Point:555 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
Frequently Bought Together