

p-Cresol 99+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-44-5
:
Formula:C₇H₈O
MDL Number:MFCD00002376
:108.14 g/mol
Merck Index:13,02604
Boiling Pt:202 °C (760 mmHg)
Melting Pt:32…34 °C
Density:1.034 g/cm³
Flash Pt:89 °C
Storage Temperature:Ambient
EINECS:203-398-6
UN:3455
ADR:6.1,II
:GHS06
:GHS05
Specification Test Results
- Appearance:Colorless to light yellow Crystalline solid or liquid (May darken on storage)
- Infrared spectrum:Conforms
- Water:≤0.05 %
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together