Order Entry
Switzerland
Orders LinkContactUsLinkComponent
m-Cresol | Avantor

m-Cresol

Glass bottle for solids, 250 g
Katalognr. ROTH9269.1
Giftig
Ätzend
Marketsource
Anbieter:   Roth Carl
m-Cresol
Katalognr. ROTH9269.1
Anbieter:  Roth Carl
Marketsource
CAS Nummer:  108-39-4
m-Cresol
m-Cresol

Glass bottle for solids, 250 g

CAS Nummer:  108-39-4
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 250 g
Product Details & Documents
Synonyme:  1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene
  • CAS Nummer:108-39-4
  • Formel:CH₃C₆H₄OH
  • MDL-Nummer:MFCD00002302
  • Molekulargewicht:108.14 g/mol
  • Merck-Index:13,02604
  • Siedepunkt:202.2 °C (1013 hPa)
  • Schmelzender Platingehalt:11.5 °C
  • Dichte:1.0336 g/cm³ (20 °C)
  • Blitz Pt:73 °C
  • Lagertemperatur:Ambient
  • EINECS:203-577-9
  • UN:2076
  • ADR:6.1,II
Technische Daten

Technische Daten

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.