

m-Cresol ≥98%, GPR RECTAPUR®
23048.261
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Assay:Min. 98 %
- Bromine water test:Passes test
- Appearance:colourless to yellowish liquid
- Ferric chloride test:Passes test
- Identification:Passes test
- Non-volatile residue:Max. 0.1 %
- p- and o-Cresol and related substances:Max. 2.0 %
:
:108.14 g/mol
:13,02604
:202.2 °C (1013 hPa)
:11.5 °C
:1.0336 g/cm³ (20 °C)
:73 °C
:Ambient
:203-577-9
:2076
:6.1 (8),II
:GHS06
:GHS05
:108-39-4
:CH₃C₆H₄OH
:MFCD00002302