Order Entry
Switzerland
ContactUsLinkComponent
m-Cresol for synthesis, Sigma-Aldrich®
Toxic
Corrosive
m-Cresol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Toxic
Corrosive
m-Cresol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    23.5 g/l (20 °C)
  • Melting Point:
    11.5 °C
  • Molar Mass:
    108.14 g/mol
  • Boiling Point:
    203 °C (1013 hPa)
  • Vapor Pressure:
    0.065 hPa (20 °C)
  • Flash Point:
    1 %(V)
  • Refractive Index:
    86 °C
  • Density:
    1.03 g/cm3 (20 °C)
  • pH:
    5 (20 g/l, H2O, 20 °C)
  • Ignition Point:
    626 °C
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together