

N,N-Dimethyl-p-toluidine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-97-8
:
Formula:C₉H₁₃N
MDL Number:MFCD00008316
:135.21 g/mol
Boiling Pt:210…211 °C
Melting Pt:–25 °C
Density:0.937
Flash Pt:83 °C (181 °F)
Storage Temperature:Ambient
EINECS:202-805-4
UN:1708
ADR:6.1,II
:GHS06
:GHS08
Related Information
Frequently Bought Together