4-Methoxybenzyl chloride
Catalog # WAKO131-05461
Supplier: Wako
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:156.61 g/mol
Boiling Pt:244 °C (1013 hPa)
Melting Pt:–1 °C
Density:1.072 g/cm³ (0 °C)
Storage Temperature:Refrigerator
EINECS:212-540-6
UN:1760
ADR:8,II
:GHS05
CAS Number:824-94-2
Formula:C₈H₉ClO
MDL Number:MFCD00000915