Order Entry
Switzerland
ContactUsLinkComponent
α-Bromo-3-iodotoulene
α-Bromo-3-iodotoulene
Catalog # MOLE39198942-5G
Supplier:  Molekula
CAS Number:  
α-Bromo-3-iodotoulene
Catalog # MOLE39198942-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1-(Bromomethyl)-3-iodobenzene, α-Bromo-m-iodotoulene, 3-Iodobenzyl bromide
 :
 :296.93 g/mol
UN:3261
 :GHS05
Storage Temperature:Ambient
CAS Number:49617-83-6
Formula:C₇H₆BrI
MDL Number:MFCD00019016
Melting Pt:50…52 °C
ADR:8 II