Order Entry
Switzerland
ContactUsLinkComponent
Pyrogallol, GPR RECTAPUR®
Health
Irritant
Pyrogallol, GPR RECTAPUR®
Catalog # 20647.237
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Pyrogallol, GPR RECTAPUR®
Catalog # 20647.237
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • IR Spectrum:
    Passes test
  • Melting point:
    131-134 °C
  • Ignition residue (SO4):
    Max. 0.1 %
  • Water:
    Max. 1 %

Specification Test Results

About this item

 :
 :126.11 g/mol
Merck Index:13,08090
Boiling Pt:309 °C (1013 hPa)
Melting Pt:130…132 °C
Density:1.453 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-762-9
UN:2811
ADR:6.1,III
 :GHS08
 :GHS07
CAS Number:87-66-1
Formula:C₆H₆O₃
MDL Number:MFCD00002192