Pyrocatechol
Plastic bottle for solids, 25 g
Catalog # ROTH4249.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
- CAS Number:120-80-9
- Formula:C₆H₆O₂
- MDL Number:MFCD00002188
- Molecular Weight:110.11 g/mol
- Merck Index:13,08089
- Boiling Pt:245 °C (1013 hPa)
- Melting Pt:103…105 °C
- Density:1.0548 g/cm³ (20 °C)
- Flash Pt:127 °C
- Storage Temperature:Ambient
- EINECS:204-427-5
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 g



