Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Pyrocatechol for synthesis, Sigma-Aldrich®

Pyrocatechol for synthesis, Sigma-Aldrich®

Supplier:   Merck
Pyrocatechol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,08089
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:103…105 °C
  • Density:1.0548 g/cm³ (20 °C)
  • Flash Pt:127 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ200
  • EINECS:204-427-5

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Solubility:
    430 g/l (20 °C)
  • Melting Point:
    103 - 105 °C
  • Molar Mass:
    110.11 g/mol
  • Boiling Point:
    246 °C (1013 hPa)
  • Vapor Pressure:
    13 hPa (118 °C)
  • Flash Point:
    127 °C
  • Explosion Limit:
    1.97 %(V)
  • Density:
    1.37 g/cm3 (20 °C)
  • pH:
    6 (100 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.