Order Entry
Switzerland
ContactUsLinkComponent
2,4-Diaminoanisole, Supelco®
Health
Irritant
Environment
2,4-Diaminoanisole, Supelco®
Catalog # 32831-100MG
Supplier:  Merck
CAS Number:  
undefined
Health
Irritant
Environment
2,4-Diaminoanisole, Supelco®
Catalog # 32831-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
Merck Index:12,03018
Boiling Pt:149…150 °C (5 mmHg)
Melting Pt:67.2 °C
UN:3077
ADR:9,III
 :M-Clarity™ quality level = MQ100
 :138.17 g/mol
EINECS:210-406-1
 :GHS08
 :GHS07
 :GHS09
CAS Number:615-05-4
Formula:C₇H₁₀N₂O