


p-Anisidine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:104-94-9
:
Formula:C₇H₉NO
MDL Number:MFCD00007864
:123.15 g/mol
Merck Index:13,00670
Boiling Pt:243 °C (1013 hPa)
Melting Pt:55…58 °C
Density:1.2 g/cm³ (20 °C)
EINECS:203-254-2
UN:3143
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:122 °C
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:21 g/l (20 °C)
- Melting Point:55 - 58 °C
- Molar Mass:123.15 g/mol
- Bulk Density:700 kg/m3
- Boiling Point:243 °C
- Vapor Pressure:0.02 hPa (20 °C)
- Flash Point:122 °C
- Density:1.06 g/cm3 (63 °C)
- pH:7.7 (1 g/l, H2O, 20 °C)
- Ignition Point:450 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
Frequently Bought Together