p-Anisidine
Catalog # EHERC10266020
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:123.15 g/mol
Merck Index:13,00670
Boiling Pt:243 °C (1013 hPa)
Melting Pt:55…58 °C
Density:1.2 g/cm³ (20 °C)
Flash Pt:122 °C
EINECS:203-254-2
UN:2431
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:104-94-9
Formula:C₇H₉NO
MDL Number:MFCD00007864