Order Entry
Switzerland
ContactUsLinkComponent
m-Anisidine
m-Anisidine
Catalog # EHERC10266010
Supplier:  EHRENSTORFER
CAS Number:  
m-Anisidine
Catalog # EHERC10266010
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :123.15 g/mol
Merck Index:13,00670
Boiling Pt:251 °C (1013 hPa)
Melting Pt:–2 °C
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-651-4
UN:2431
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:536-90-3
Formula:C₇H₉NO
MDL Number:MFCD00007783