m-Anisidine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:536-90-3
:
Formula:C₇H₉NO
MDL Number:MFCD00007783
:123.15 g/mol
Merck Index:13,00670
Boiling Pt:250…251 °C (760 mmHg)
Melting Pt:-2 °C
Density:1.102 g/cm³
Flash Pt:126 °C
Storage Temperature:Ambient
EINECS:208-651-4
UN:2431
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5800 to 1.5820 (20°C, 589 nm)
- GC:≥98.5 %
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together