o-Anisidine
Catalog # EHERC10266000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:123.15 g/mol
Merck Index:13,00670
Boiling Pt:225 °C (1013 hPa)
Melting Pt:5 °C
Density:1.09 g/cm³ (20 °C)
Flash Pt:118 °C
Storage Temperature:Ambient
EINECS:201-963-1
UN:2431
ADR:6.1,III
:GHS06
:GHS08
CAS Number:90-04-0
Formula:C₇H₉NO
MDL Number:MFCD00007688