Order Entry
Switzerland
ContactUsLinkComponent
3,5-Dinitroaniline 97%, Sigma-Aldrich®
Toxic
Health
Environment
3,5-Dinitroaniline 97%, Sigma-Aldrich®
Catalog # D193402-10G
CAS Number:  
undefined
Toxic
Health
Environment
3,5-Dinitroaniline 97%, Sigma-Aldrich®
Catalog # D193402-10G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance (Colour):
    Faint Yellow to Dark Yellow and Faint Brown to Dark Brown
  • Appearance (Form):
    Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    38.0 - 40.7 %
  • Nitrogen:
    22.1 - 23.7 %
  • Purity (GC):
    ≥96.5 %

Specification Test Results

About this item

 :
Melting Pt:160...162 °C (lit.)
Density:1.601 g/cm³ (20 °C)
EINECS:210-567-8
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
 :183.12 g/mol
UN:1596
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Room Temperature
CAS Number:618-87-1
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007263