

N,N-Dimethyl-4-nitrosoaniline 97%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:138-89-6
:
Formula:C₈H₁₀N₂O
MDL Number:MFCD00002063
:150.18 g/mol
Merck Index:12,06736
Boiling Pt:259 °C (1013 hPa)
Melting Pt:85...87 °C (lit.)
Density:1.145 g/cm³ (20 °C)
UN:1369
ADR:4.2,II
:M-Clarity™ quality level = MQ100
EINECS:205-343-1
:GHS02
:GHS06
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Form):Powder and/or Chunks
Conforms to Requirements - Infrared spectrum:Conforms to Structure
- Carbon:61.7 - 66.2 %
- Nitrogen:18.0 - 19.3 %
- Purity (TLC):≥97 %
- Solubility (Turbidity):Clear to Turbid
- Solubility (Colour)
c = 5%, Ethanol:Green to Very Dark Green
Specification Test Results
Frequently Bought Together