Order Entry
Switzerland
ContactUsLinkComponent
 
Pentafluorobenzoyl chloride ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
 :  Merck
 
Pentafluorobenzoyl chloride ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Assay (GC, area%):
    ≥98.0%
  • Identity (IR):
    Passes test
  • Boiling Point:
    158 - 159 °C (1013 hPa)
  • Density:
    1.664 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) acidic

 

Frequently Bought Together