Order Entry
Switzerland
ContactUsLinkComponent
Methyl salicylate ≥99%, GPR RECTAPUR®
Irritant
Methyl salicylate ≥99%, GPR RECTAPUR®
Catalog # 25616.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Methyl salicylate ≥99%, GPR RECTAPUR®
Catalog # 25616.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Density (20/4):
    1.180 - 1.190

Specification Test Results

About this item

 :
 :152.15 g/mol
Merck Index:13,06143
Boiling Pt:432 °C (1013 hPa)
Melting Pt:100…101 °C
Density:1.184 g/cm³ (20 °C)
Flash Pt:99 °C
Storage Temperature:Ambient
EINECS:204-317-7
 :GHS07
CAS Number:119-36-8
Formula:2-(HO)C₆H₄CO₂CH₃
MDL Number:MFCD00002214