Ethyl-4-nitrobenzoate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-77-4
:
Formula:C₉H₉NO₄
MDL Number:MFCD00007351
Melting Pt:56…59 °C
:195.17 g/mol
Storage Temperature:Ambient
EINECS:202-786-2
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:56 to 59 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together