ContactUsLinkComponent
Membrane dispenser for microbiological control, electronic butler (eButler), Whatman™
Membrane dispenser for microbiological control, electronic butler (eButler), Whatman™
Catalog # WHAT10477103
undefined
Membrane dispenser for microbiological control, electronic butler (eButler), Whatman™
Catalog # WHAT10477103
Specifications
  • Description:
    Electronic butler (eButler) membrane dispenser
  • Pk:
    1
Specifications
About this item
The eButler rapid membrane dispenser allows for more efficient microbiological testing. It rapidly dispenses culture membranes using either hands-free or push-button operation.
  • Aluminum and stainless steel construction
  • Designed for easy membrane loading and device usability
  • Up to 400 membranes per package
  • Compatible with Whatman™ sterile mixed cellulose ester (MCE) microbiology membranes
This is the only eButler compatible with Whatman™ MCE microbiology membranes.