Volumetric flasks, class A, blue markings, BLAUBRAND®
Nome del fornitore: Brand
Certificati
Informazioni su questo articolo
Borosilicate glass 3,3, with beaded rim.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
* Wide neck
Certifications: DIN EN ISO 1042
Ordering information: Supplied with an individual certificate.
Frequently Bought Together