Order Entry
Switzerland
ContactUsLinkComponent
2,3,5,6-Tetrachlorophenol analytical standard, Supelco®
2,3,5,6-Tetrachlorophenol analytical standard, Supelco®
Catalog # 442284.
Supplier:  Merck
CAS Number:  
undefined
2,3,5,6-Tetrachlorophenol analytical standard, Supelco®
Catalog # 442284.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    0001

Specifications

About this item

 :
 :231.89 g/mol
Boiling Pt:261 °C (1013 hPa)
Melting Pt:114…115 °C
Density:1.6 g/cm³ (20 °C)
EINECS:213-310-8
UN:2020
ADR:6.1,III
CAS Number:935-95-5
Formula:C₆H₂Cl₄O
MDL Number:MFCD00044671