

2-Chlorophenyl isothiocyanate 99%
Catalog # ACRO292240050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6598 to 1.6630 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
About this item
:
:169.63 g/mol
Boiling Pt:260 °C (1013 hPa)
Density:1.29 g/cm³
Flash Pt:>110 °C
UN:2810
ADR:6.1,II
:GHS06
:GHS08
Storage Temperature:Refrigerator
CAS Number:2740-81-0
Formula:C₇H₄ClNS
MDL Number:MFCD00004801