Powder funnels, stainless steel
Anbieter: Sampling Systems
Zertifikate
Über diesen Artikel
These powder funnels are made from hygienic 316 stainless steel.
- Satin finish
- 2" tri ferrule fitting/opening at spout base
- Very good chemical and temperature resistance
The powder funnel has a wide 50 mm diameter outlet to help prevent blackages. The outlet has a 2" ferrule so the the powder funnel can be clamped to a suitable branch.
Frequently Bought Together