Chlorobenzene for HPLC
:108-90-7
:
:C₆H₅Cl
:MFCD00000530
:112.56 g/mol
:13,02139
:130 °C (1013 hPa)
:–45 °C
:1.11 g/cm³ (20 °C)
:28 °C
:Ambient
:203-628-5
:1134
:3,III
:01-2119432722-45
:GHS02
:GHS07
:GHS09
Frequently Bought Together