2-Bromophenol for synthesis, Sigma-Aldrich®
8.04166.0025
: Merck
:
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
- Solubility:(20 °C) practically insoluble
- Melting Point:5 °C
- Molar Mass:173.01 g/mol
- Boiling Point:195 °C
- Flash Point:108 °C
- Refractive Index:1.5893 (20 °C, 589 nm)
- Density:1.63 g/cm3 (20 °C)
:
:173.01 g/mol
:13,01413
:195 °C (1013 hPa)
:5.6 °C
:1.49 g/cm³ (20 °C)
:Ambient
:202-432-7
:3082
:9,III
:M-Clarity™ quality level = MQ200
:GHS09
:95-56-7
:C₆H₅BrO
:MFCD00002146