ContactUsLinkComponent
WiFi streamer, Dino-Lite WF-10
WiFi streamer, Dino-Lite WF-10
  630-2923
undefined
WiFi streamer, Dino-Lite WF-10
  630-2923
  • Description:
    Wi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion battery
  • Seq:
    901
  • Pk:
    1
WF-10 allows compatible Dino-Lite USB microscopes to be used wirelessly in connection with an iOS (iPhone/iPad) or Android app on any tablet, smartphone, or even in the DinoCapture software for Windows computers.
  • Can be used anywhere to create a WiFi signal for streaming the Dino-Lite live images
  • Can stream from one Dino-Lite microscope to multiple devices
  • Built-in battery is replaceable
Ideal for field work or presentations.
Compatible with: AM/AD4xxx (R4), AM4xx5, AM/AD7xxx (R4), MEDL4xx (R4), MEDL7xx (R4) series.
Please note: The free DinoConnect App needs to be downloaded from the App Store or Google Play.