Order Entry
Switzerland
ContactUsLinkComponent
1-Bromo-4-fluorobenzene analytical standard, Supelco®
1-Bromo-4-fluorobenzene analytical standard, Supelco®
Catalog # 442404.
Supplier:  Merck
CAS Number:  
undefined
1-Bromo-4-fluorobenzene analytical standard, Supelco®
Catalog # 442404.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Boiling Pt:151 °C (1013 hPa)
Melting Pt:–16 °C
Density:1.4946 g/cm³ (20 °C)
Flash Pt:53 °C
Storage Temperature:Ambient
EINECS:207-300-2
UN:1993
ADR:3,III
 :175 g/mol
CAS Number:460-00-4
Formula:C₆H₄BrF
MDL Number:MFCD00000342