



n-Pentane ≥99.5% (GC, on anhydrous basis), PESTINORM® Supra Trace
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:109-66-0
:
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
:72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under inert gas.
Flash Pt:–40 °C
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear colorless liquid
- Assay (GC, on anhydrous basis):min. 99.5 %
- Acidity (as Acetic acid):max. 0.002 %
- Color (APHA, Pt-Co):max. 5
- Residue after evaporation:max. 0.0001 % w/w
- Water (KF):max. 0.005 % w/w
- GC/ECD Dioxins, Furans & PCB's:max. 5 pg/ml
- GC/ECD any Pesticide (as Lindane):max. 2 pg/ml
- GC/NPD any Pesticide (as Parathion):max. 5 pg/ml
- GC/ECD 1,2,4-TCB to daca-PCB (as Lindane):max. 5 pg/ml
- GC/ECD DCM to 1,2,4-TCB (as TCM):max. 2 ng/ml
- GC/FID C10 to C40 (as n-Decane):max. 2 ng/ml
- GC/MSD C10 to C40 (as Decane; 30-600 amu):max. 2 ng/ml
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_PESTINORM® SUPRATRACE n-Pentane 1 l spec_PESTINORM® SUPRATRACE n-Pentane 2,5 l
Product specification
VWR® PESTINORM® Solvents
Product Literature
Frequently Bought Together