Benzimidazole 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51-17-2
:
Formula:C₇H₆N₂
MDL Number:MFCD00005585
:118.14 g/mol
Merck Index:13,01082
Boiling Pt:360 °C
Melting Pt:171…174 °C
Density:1,23 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-081-4
Specification Test Results
- Appearance:Beige to brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:171 to 174 °C
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together