Xylene (mixture of isomers) for histology
Catalog # 1.08298.4000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
Specifications
About this item
:
:106.17 g/mol
Boiling Pt:136…145 °C (1013 hPa)
Melting Pt:–25 °C
Storage Temperature:Ambient
UN:1307
ADR:3,III
:GHS02
:GHS08
:GHS07
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264